Assay Details
Gene Symbol: Rpp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025938.4 | NP_080214.1 | 3 - 4 | 371 | 100 | |
GenBank mRNA | AK012105.1 | - | 3 - 4 | 318 | 100 | |
AK016631.1 | - | 3 - 4 | 318 | 100 | ||
BC026988.1 | - | 3 - 4 | 301 | 5368565 | 100 | |
BC046803.1 | - | 3 - 4 | 299 | 6404792 | 100 | |
CF896465.1 | - | 3 - 4 | 371 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29387669 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29387668 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29387666 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29387667 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P10433 | BC046803.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P10433 | AK016631.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P10433 | BC026988.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P10433 | NM_025938.4 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P10433 | AK012105.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 550027 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 348641 | AB006960.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 348641 | AK143657.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 348641 | BC053025.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 348641 | NM_016678.1 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 719193 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 204018 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 606730 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 915632 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 486287 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 648881 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 944075 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 59726 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P105970 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P138685 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P10433 | BC046803.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P10433 | AK016631.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P10433 | BC026988.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P10433 | NM_025938.4 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P10433 | AK012105.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P167273 | AK016631.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29387640 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29387645 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29387666 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29387669 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29387657 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29387643 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29387670 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29387663 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29387678 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29387677 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29387649 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29387667 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29387672 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29387650 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29387655 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29387662 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29387653 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29387658 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29387660 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29387676 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29387654 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29387671 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P105970 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P138685 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08021585 | - | - |
Whole Mouse Genome G4122A | A_51_P167273 | AK016631.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125326 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P105970 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P138685 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11711508 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11825503 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11711510 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11711512 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29387666 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29387667 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29387669 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29387668 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387745 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387746 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387703 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387670 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387759 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387713 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387750 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387747 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387662 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387770 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387757 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387781 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387666 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387740 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387672 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387777 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387682 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387742 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387761 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387664 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387692 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387643 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387700 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387732 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387667 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387719 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387659 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387779 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387694 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387712 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387722 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387685 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387720 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387707 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387684 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387710 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387753 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387714 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387651 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387635 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387734 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387773 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387762 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387772 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387775 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387653 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387655 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387690 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387711 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387767 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387755 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387669 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387718 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387730 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387735 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387657 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387679 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387645 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387634 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387763 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387738 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387729 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387765 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387737 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387676 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387649 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387687 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387774 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387727 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387660 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387708 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29387671 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29387637 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29387645 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11711513 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29387655 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29387687 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29387699 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29387697 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11142532 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29387718 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29387651 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29387696 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29387704 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29387677 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11711509 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11142529 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29387664 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29387700 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11628407 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29387667 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29387678 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29387635 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29387684 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29387653 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29387640 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11734558 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29387643 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11734559 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11760031 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29387666 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11821218 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29387662 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11711515 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29387649 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11196813 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29387654 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29387685 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29387682 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11647240 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11629989 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29387676 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29387674 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29387692 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29387657 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29387690 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29387679 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10412623 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10412618 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10412622 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10412621 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10412617 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10412620 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10412619 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1419460_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1419461_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10412616 | - | - |
Mouse Genome 430 2.0 Array | 1419460_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1419460_at | - | - |
Mouse Genome 430 2.0 Array | 1419461_at | - | - |
Mouse Genome 430 2.0 Array | 1419461_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67053_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67053_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1400000679.mm.1 | NM_025938.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400000679.mm.1 | NM_025938.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400000679.mm.1 | TC1400000075.mm.1 | MTA TC |
Mouse Expression Set 430 | 1419460_PM_at | - | - |
Mouse Expression Set 430 | 1419460_at | - | - |
Mouse Expression Set 430 | 1419461_at | - | - |
Mouse Expression Set 430 | 1419461_PM_at | - | - |
Murine Genome U74v2 Set | 99182_at | - | - |
Murine Genome U74v2 Set | 160474_at | - | - |
Murine Genome U74v2 Set | 99182_PM_at | - | - |
Murine Genome U74v2 Set | 160474_PM_at | - | - |
Murine Genome U74v2 Set | 168317_r_at | - | - |
Murine Genome U74v2 Set | 168317_PM_r_at | - | - |
Murine Genome U74v2 Set | 168318_PM_at | - | - |
Murine Genome U74v2 Set | 168318_at | - | - |
Mouse Genome 430A 2.0 Array | 1419460_at | - | - |
Mouse Genome 430A 2.0 Array | 1419461_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1419460_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1419461_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1419460_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1419460_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1419461_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1419461_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1443358_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1443358_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 299116 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 7040014 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 7040014 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0007040014 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1218584 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1218584 | - | - |
mouse-6 v1.1 (Illumina) | scl46641.9_62-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 7040014 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl46641.9_62-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 7040014 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1218584 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1218584 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl46641.9_62-S | - | - |
