Assay Details
Gene Symbol: Trappc6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025960.3 | NP_080236.2 | 4 - 5 | 381 | 115 | |
GenBank mRNA | AK007965.1 | - | 4 - 5 | 380 | 115 | |
AK008022.1 | - | 4 - 5 | 381 | 115 | ||
AK008462.2 | - | 4 - 5 | 380 | 115 | ||
AK088832.1 | - | 4 - 5 | 380 | 115 | ||
BC020128.1 | - | 3 - 4 | 330 | 115 | ||
BC037154.1 | - | 4 - 5 | 418 | 2811950 | 115 | |
BC106114.1 | - | 4 - 5 | 395 | 4168174 | 115 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
