Assay Details
| Gene Symbol: Mmachc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025962.3 | NP_080238.2 | 1 - 2 | 149 | 65 | |
| GenBank mRNA | AK007725.1 | - | 1 - 2 | 84 | 65 | |
| AK012761.1 | - | 1 - 2 | 86 | 65 | ||
| AK084194.1 | - | 1 - 2 | 100 | 65 | ||
| BB614283.1 | - | 1 - 2 | 150 | 65 | ||
| BC054756.1 | - | 1 - 2 | 88 | 6411797 | 65 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK007725.1,AK084194.1,BC054756.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P388298 | AK084194.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P388298 | BC054756.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P388298 | NM_025962.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P388298 | AK007725.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P388298 | AK084194.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P388298 | BC054756.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P388298 | NM_025962.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P388298 | AK007725.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 318640 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 307835 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 706561 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 64991 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P141513 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P388298 | AK084194.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P388298 | BC054756.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P388298 | NM_025962.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P388298 | AK007725.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P388298 | AK084194.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P388298 | BC054756.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P388298 | NM_025962.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P388298 | AK007725.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23230548 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23230534 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23230557 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23230543 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23230542 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23230533 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23230550 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23230560 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23230535 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23230558 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23230559 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23230538 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23230565 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23230563 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23230536 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23230540 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23230553 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23230549 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23230546 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23230556 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23230555 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P141513 | - | - |
| Whole Mouse Genome G4122A | A_51_P388298 | AK084194.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P388298 | BC054756.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P388298 | NM_025962.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P388298 | AK007725.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P388298 | AK084194.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P388298 | BC054756.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P388298 | NM_025962.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P388298 | AK007725.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135943 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P141513 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03879473 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03879481 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03879482 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03879486 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03879485 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03879475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23230548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23230599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23230557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23230481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23230478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23230475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23230540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23230472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23230555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23230546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23230523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23230550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23230544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23230493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23230541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23230492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23230477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23230545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23230553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23230560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23230565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23230563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23230558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23230482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23230487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23230532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23230489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23230526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23230536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23230559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23230535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23230549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23230530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23230547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23230483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23230488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23230528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23230556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23230598 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03972377 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23230540 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23230550 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23230535 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03879470 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23230523 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23230532 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23230553 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23230544 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03851717 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03966898 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23230559 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23230558 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23230537 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23230555 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23230533 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23230551 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23230545 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23230534 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23230526 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23230560 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23230530 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23230542 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03725056 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23230541 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23230528 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03966899 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23230543 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03208539 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10515366 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515367 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515365 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515364 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418915_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10515363 | - | - |
| Mouse Genome 430 2.0 Array | 1418915_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1418915_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67096_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67096_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400026773.mm.1 | TC0400003349.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400026773.mm.1 | NM_025962.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400026773.mm.1 | NM_025962.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400026774.mm.1 | TC0400003349.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400026774.mm.1 | NM_025962.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400026774.mm.1 | NM_025962.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400026776.mm.1 | TC0400003349.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400026776.mm.1 | NM_025962.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400026776.mm.1 | NM_025962.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400026777.mm.1 | TC0400003349.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400026777.mm.1 | NM_025962.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400026777.mm.1 | NM_025962.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400026778.mm.1 | TC0400003349.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400026778.mm.1 | NM_025962.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400026778.mm.1 | NM_025962.3 | Refseq |
| Mouse Expression Set 430 | 1418915_PM_at | - | - |
| Mouse Expression Set 430 | 1418915_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418915_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418915_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418915_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418915_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 753573 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6380176 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6380176 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 102320731 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006380176 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1221958 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2446992 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1221958 | - | - |
| mouse-6 v1.1 (Illumina) | scl0067096.2_233-S | - | - |
| mouse-6 v1.1 (Illumina) | scl23949.4_43-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 102320731 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6380176 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0067096.2_233-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6380176 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102320731 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2446990 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2446990 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0067096.2_233-S | - | - |

Mmachc
Genomic Map