Assay Details
| Gene Symbol: Ptgr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025968.3 | NP_080244.1 | 4 - 5 | 259 | 86 | |
| GenBank mRNA | AK010888.1 | - | 4 - 5 | 294 | 86 | |
| AK011962.1 | - | 4 - 5 | 281 | 86 | ||
| AK035425.1 | - | 4 - 5 | 260 | 86 | ||
| AK134440.1 | - | 4 - 5 | 292 | 86 | ||
| AK166835.1 | - | 4 - 5 | 259 | 86 | ||
| BC014865.1 | - | 4 - 5 | 241 | 2648051 | 86 | |
| CT010267.1 | - | 3 - 4 | 208 | 86 | ||
More Information
Set Membership: |
|

Ptgr1
Genomic Map