Assay Details
| Gene Symbol: Mastl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025979.4 | NP_080255.3 | 9 - 10 | 2384 | 62 | |
| GenBank mRNA | AK006448.1 | - | 4 - 5 | 1485 | 62 | |
| AK012597.1 | - | 4 - 5 | 1485 | 62 | ||
| AK030140.1 | - | 9 - 10 | 2279 | 62 | ||
| BC066834.1 | - | 1641 | 62 | |||
| BC086483.1 | - | 1653 | 62 | |||
| BC103779.1 | - | 9 - 10 | 2260 | 30480071 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mastl
Genomic Map