Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400028490.mm.1 |
TC0400003506.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400028490.mm.1 |
NM_026031.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400028490.mm.1 |
NM_026031.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400028492.mm.1 |
TC0400003506.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400028492.mm.1 |
NM_026031.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400028492.mm.1 |
NM_026031.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400028493.mm.1 |
TC0400003506.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400028493.mm.1 |
NM_026031.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400028493.mm.1 |
NM_026031.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400028495.mm.1 |
TC0400003506.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400028488.mm.1 |
TC0400003506.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400028488.mm.1 |
NM_026031.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400028488.mm.1 |
NM_026031.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400028491.mm.1 |
TC0400003506.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400028491.mm.1 |
NM_026031.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400028491.mm.1 |
NM_026031.3 |
Refseq |