Assay Details
| Gene Symbol: Ndufb8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026061.2 | NP_080337.1 | 4 - 5 | 486 | 71 | |
| GenBank mRNA | AK010177.1 | - | 4 - 5 | 476 | 71 | |
| AK013516.1 | - | 4 - 5 | 486 | 71 | ||
| BC043019.1 | - | 4 - 5 | 486 | 5684256 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32132881 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32132941 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32132940 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32132943 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32132882 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32132942 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32132884 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32132966 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32132939 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32132945 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32132944 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P247873 | BC043019.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P247873 | AK013516.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P247873 | NM_026061.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P247873 | AK010177.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P247873 | BC043019.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P247873 | AK013516.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P247873 | NM_026061.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P247873 | AK010177.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 35323 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 580752 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 676282 | AK049238.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 676282 | BC026912.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 676282 | NM_001013616.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 676282 | XM_921499.3 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 48708 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P118831 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P247873 | BC043019.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P247873 | AK013516.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P247873 | NM_026061.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P247873 | AK010177.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P247873 | BC043019.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P247873 | AK013516.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P247873 | NM_026061.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P247873 | AK010177.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P252539 | AK028070.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P247873 | BC043019.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P247873 | AK013516.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P247873 | NM_026061.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P247873 | AK010177.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P247873 | BC043019.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P247873 | AK013516.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P247873 | NM_026061.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P247873 | AK010177.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32132933 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32132940 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32132953 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32132935 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32132929 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32132949 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32132951 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32132943 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32132964 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32132961 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32132941 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32132960 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32132965 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32132948 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32132938 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32132928 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32132936 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32132932 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32132966 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32132958 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32132946 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32132942 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32132939 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32132956 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32132931 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32132944 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P118831 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03603441 | - | - |
| Whole Mouse Genome G4122A | A_52_P252539 | AK028070.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P247873 | BC043019.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P247873 | AK013516.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P247873 | NM_026061.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P247873 | AK010177.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P247873 | BC043019.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P247873 | AK013516.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P247873 | NM_026061.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P247873 | AK010177.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P126568 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P118831 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14393455 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14474442 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14393445 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14393450 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14474443 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14393456 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14498008 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14393451 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32132940 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32132943 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32132884 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32132941 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32132939 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32132942 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32132944 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32132966 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32132881 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32132882 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32132945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32132948 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14447238 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14393451 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32132933 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32132951 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32132915 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14393460 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14393442 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32132948 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14447237 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32132910 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32132949 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14212559 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32132918 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32132917 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32132923 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32132943 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14486243 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32132958 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32132940 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14312321 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32132932 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32132944 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32132936 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32132931 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32132926 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32132953 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32132914 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32132939 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32132928 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14428683 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32132938 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32132946 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10468017 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468019 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468020 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468018 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468021 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1458404_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448198_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10468016 | - | - |
| Mouse Genome 430 2.0 Array | 1448198_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1458404_at | - | - |
| Mouse Genome 430 2.0 Array | 1458404_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1448198_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67264_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67264_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900013877.mm.1 | NM_026061.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900013877.mm.1 | NM_026061.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900013877.mm.1 | TC1900001777.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1448198_PM_a_at | - | - |
| Mouse Expression Set 430 | 1448198_a_at | - | - |
| Mouse Expression Set 430 | 1458404_at | - | - |
| Mouse Expression Set 430 | 1458404_PM_at | - | - |
| Murine Genome U74v2 Set | 93581_PM_at | - | - |
| Murine Genome U74v2 Set | 93581_at | - | - |
| Murine Genome U74v2 Set | 170210_PM_i_at | - | - |
| Murine Genome U74v2 Set | 170210_i_at | - | - |
| Murine Genome U74v2 Set | 165726_PM_at | - | - |
| Murine Genome U74v2 Set | 165726_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448198_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448198_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448198_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448198_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1458404_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1458404_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 352315 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2260450 | - | - |
| MouseWG-6 v2 Expression BeadChip | 105670041 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 105670041 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2260450 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002260450 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2621966 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2621966 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2501071 | - | - |
| mouse-6 v1.1 (Illumina) | scl0067264.2_318-S | - | - |
| mouse-6 v1.1 (Illumina) | scl52441.8_4-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 105670041 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2260450 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0067264.2_318-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 105670041 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2260450 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2621966 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2621966 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2501071 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0067264.2_318-S | - | - |

Ndufb8
Genomic Map