Assay Details
| Gene Symbol: Pdlim7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114087.2 | NP_001107559.1 | 1 - 2 | 82 | 66 | |
| NM_001114088.2 | NP_001107560.1 | 1 - 2 | 82 | 66 | ||
| NM_026131.4 | NP_080407.3 | 1 - 2 | 82 | 66 | ||
| NR_104287.1 | 1 - 2 | 82 | 66 | |||
| GenBank mRNA | AK003338.1 | - | 1 - 2 | 73 | 66 | |
| AK010141.1 | - | 1 - 2 | 71 | 66 | ||
| AK010339.1 | - | 1 - 2 | 73 | 66 | ||
| AK033091.1 | - | 74 | 66 | |||
| AK049902.1 | - | 1 - 2 | 74 | 66 | ||
| AK077998.1 | - | 1 - 2 | 74 | 66 | ||
| AK128987.1 | - | 1 - 2 | 70 | 66 | ||
| AK142933.1 | - | 1 - 2 | 64 | 66 | ||
| AK167476.1 | - | 1 - 2 | 74 | 66 | ||
| BC049565.1 | - | 1 - 2 | 70 | 6588160 | 66 | |
| BC052698.1 | - | 1 - 2 | 53 | 66 | ||
| BY056634.1 | - | 1 - 2 | 75 | 66 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK003338.1,AK010141.1,AK077998.1,AK167476.1,BC049565.1,BC052698.1,NM_001114087.2,NM_001114088.2,NM_026131.4 transcripts.
Set Membership: |
|

Pdlim7
Genomic Map