Assay Details
| Gene Symbol: Cbl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007619.2 | NP_031645.2 | 14 - 15 | 2430 | 63 | |
| GenBank mRNA | AK029826.1 | - | 14 - 15 | 2433 | 63 | |
| AK153915.1 | - | 14 - 15 | 2382 | 63 | ||
| BC125285.1 | - | 14 - 15 | 2345 | 40129800 | 63 | |
| X57111.1 | - | 14 - 15 | 2317 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cbl
Genomic Map