Assay Details
| Gene Symbol: Cbr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007621.2 | NP_031647.1 | 2 - 3 | 204 | 70 | |
| GenBank mRNA | AK081816.1 | - | 2 - 3 | 175 | 70 | |
| BC010758.1 | - | 2 - 3 | 164 | 4219349 | 70 | |
| BY118518.1 | - | 2 - 3 | 205 | 70 | ||
| D26123.1 | - | 2 - 3 | 191 | 70 | ||
| X07411.1 | - | 2 - 3 | 161 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2122605 | AK081816.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2122605 | BC010758.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2122605 | BF730701.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2122605 | D26123.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2122605 | X07411.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2122604 | AK081816.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2122604 | BC010758.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2122604 | D26123.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2122604 | X07411.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2122604 | NM_007621.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 200209 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2122604 | AK081816.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2122604 | BC010758.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2122604 | D26123.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2122604 | X07411.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2122604 | NM_007621.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2122605 | AK081816.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2122605 | BC010758.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2122605 | BF730701.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2122605 | D26123.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2122605 | X07411.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P238425 | AK081816.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P238425 | BC010758.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P238425 | BF730701.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P238425 | D26123.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P238425 | X07411.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186478 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186468 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186462 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186467 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186463 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186470 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186465 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186473 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186481 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186494 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186487 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186479 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186475 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186489 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186483 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186488 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186472 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186453 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186454 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186492 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186485 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186477 | - | - |
| Whole Mouse Genome G4122A | A_51_P238425 | AK081816.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P238425 | BC010758.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P238425 | BF730701.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P238425 | D26123.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P238425 | X07411.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125636 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09623069 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09623077 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09623064 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09623060 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09623062 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09623068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186475 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28186494 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28186453 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28186472 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28186454 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28186488 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28186481 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09383031 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28186460 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09171190 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09441191 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28186470 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28186492 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28186485 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09623060 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28186483 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28186489 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09276161 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09383025 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09383011 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09623064 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28186475 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28186477 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28186462 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09623068 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28186496 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28186467 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10393940 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393937 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393939 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393943 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393941 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393942 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393938 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418509_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10393936 | - | - |
| Mouse Genome 430 2.0 Array | 1418509_at | - | - |
| Mouse Genome 430 2.0 Array | 1418509_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12409_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12409_PM_at | - | - |
| Affymetrix Murine Genome 6500A Array | Msa15350_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100040474.mm.1 | NM_007621.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100040474.mm.1 | NM_007621.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100040474.mm.1 | TC1100004229.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100040473.mm.1 | NM_007621.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100040473.mm.1 | NM_007621.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100040473.mm.1 | TC1100004229.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100040477.mm.1 | NM_007621.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100040477.mm.1 | NM_007621.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100040477.mm.1 | TC1100004229.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100040478.mm.1 | NM_007621.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100040478.mm.1 | NM_007621.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100040478.mm.1 | TC1100004229.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1418509_PM_at | - | - |
| Mouse Expression Set 430 | 1418509_at | - | - |
| Murine Genome U74v2 Set | 93808_at | - | - |
| Murine Genome U74v2 Set | 93808_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418509_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418509_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418509_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418509_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 741060 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3800102 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003800102 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2756667 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2756667 | - | - |
| mouse-6 v1.1 (Illumina) | scl39222.8.1_55-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3800102 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl39222.8.1_55-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3800102 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2756665 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2756665 | - | - |

Cbr2
Genomic Map