Assay Details
| Gene Symbol: Cct5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007637.2 | NP_031663.1 | 9 - 10 | 1419 | 69 | |
| GenBank mRNA | AK077753.1 | - | 9 - 10 | 1378 | 69 | |
| AK088184.1 | - | 9 - 10 | 1353 | 69 | ||
| AK129056.1 | - | 9 - 10 | 1325 | 69 | ||
| AK133857.1 | - | 9 - 10 | 1377 | 69 | ||
| AK136493.1 | - | 9 - 10 | 1371 | 69 | ||
| AK145445.1 | - | 9 - 10 | 1388 | 69 | ||
| AK146405.1 | - | 9 - 10 | 1391 | 69 | ||
| AK146425.1 | - | 9 - 10 | 1371 | 69 | ||
| AK150390.1 | - | 9 - 10 | 1338 | 69 | ||
| AK153911.1 | - | 9 - 10 | 1395 | 69 | ||
| AK167895.1 | - | 9 - 10 | 1360 | 69 | ||
| AK213471.1 | - | 1 - 2 | 93 | 69 | ||
| BC092233.1 | - | 9 - 10 | 1389 | 69 | ||
| BC094427.1 | - | 9 - 10 | 1366 | 30537966 | 69 | |
| Z31555.1 | - | 9 - 10 | 1419 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cct5
Genomic Map