Assay Details
Gene Symbol: Cdc25a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007658.3 | NP_031684.3 | 13 - 14 | 1706 | 73 | |
GenBank mRNA | AK077609.1 | - | 13 - 14 | 1324 | 73 | |
AK077991.1 | - | 13 - 14 | 1708 | 73 | ||
AK142334.1 | - | 12 - 13 | 1981 | 73 | ||
BC046296.1 | - | 13 - 14 | 1325 | 6486343 | 73 | |
U27323.1 | - | 13 - 14 | 1295 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
