Assay Details
Gene Symbol: Coch | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198835.1 | NP_001185764.1 | 7 - 8 | 573 | 99 | |
NM_007728.5 | NP_031754.1 | 6 - 7 | 772 | 99 | ||
GenBank mRNA | AF006741.1 | - | 7 - 8 | 557 | 99 | |
AK028690.1 | - | 6 - 7 | 659 | 99 | ||
AK137336.1 | - | 6 - 7 | 774 | 99 | ||
AK156430.1 | - | 7 - 8 | 542 | 99 | ||
AK171875.1 | - | 7 - 8 | 542 | 99 | ||
BC045137.1 | - | 7 - 8 | 524 | 6306601 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
