Assay Details
| Gene Symbol: Crat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007760.3 | NP_031786.2 | 4 - 5 | 877 | 115 | |
| GenBank mRNA | AK041431.1 | - | 4 - 5 | 652 | 115 | |
| AK132179.1 | - | 5 - 6 | 704 | 115 | ||
| AK170740.1 | - | 4 - 5 | 663 | 115 | ||
| BC006668.1 | - | 4 - 5 | 624 | 3499169 | 115 | |
| X85983.1 | - | 4 - 5 | 595 | 115 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Crat
Genomic Map