Assay Details
| Gene Symbol: Ganab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293621.1 | NP_001280550.1 | 4 - 5 | 445 | 68 | |
| NM_008060.2 | NP_032086.1 | 4 - 5 | 445 | 68 | ||
| GenBank mRNA | AK028655.1 | - | 4 - 5 | 413 | 68 | |
| AK030722.1 | - | 4 - 5 | 414 | 68 | ||
| AK081915.1 | - | 4 - 5 | 407 | 68 | ||
| AK149686.1 | - | 4 - 5 | 402 | 68 | ||
| AK166150.1 | - | 4 - 5 | 384 | 68 | ||
| BC094437.1 | - | 4 - 5 | 395 | 30682580 | 68 | |
| BC117888.1 | - | 4 - 5 | 388 | 40096413 | 68 | |
| BC117889.1 | - | 4 - 5 | 391 | 40096414 | 68 | |
| BC128069.1 | - | 4 - 5 | 389 | 40096428 | 68 | |
| U92793.1 | - | 4 - 5 | 391 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
carbohydrate metabolic process
N-glycan processing
metabolic process
catalytic activity
hydrolase activity, hydrolyzing O-glycosyl compounds
protein binding
glucosidase activity
hydrolase activity
hydrolase activity, acting on glycosyl bonds
carbohydrate binding
glucan 1,3-alpha-glucosidase activity
poly(A) RNA binding
carbohydrate metabolic process
N-glycan processing
metabolic process
catalytic activity
hydrolase activity, hydrolyzing O-glycosyl compounds
protein binding
glucosidase activity
hydrolase activity
hydrolase activity, acting on glycosyl bonds
carbohydrate binding
glucan 1,3-alpha-glucosidase activity
poly(A) RNA binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2048209 | U92793.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2048209 | BC035423.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2048209 | BC094437.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2048209 | BC020386.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2048209 | AK166150.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2048210 | U92793.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2048210 | BC035423.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2048210 | BC094437.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2048210 | BC020386.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2048210 | AK166150.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 265778 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 590241 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 272561 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 650867 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 17106 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 282710 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 311311 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 810861 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 627560 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 48774 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 113900 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 91522 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P145078 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P130896 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2048209 | U92793.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2048209 | BC035423.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2048209 | BC094437.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2048209 | BC020386.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2048209 | AK166150.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2048210 | U92793.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2048210 | BC035423.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2048210 | BC094437.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2048210 | BC020386.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2048210 | AK166150.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P291936 | U92793.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P291936 | BC035423.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P291936 | BC094437.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P291936 | BC020386.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P291936 | AK214911.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P594215 | BC128069.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P594215 | U92793.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P594215 | BC117888.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P594215 | BC117889.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P594215 | BC094437.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P291936 | U92793.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P291936 | BC035423.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P291936 | BC094437.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P291936 | BC020386.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P291936 | AK214911.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123316 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110285 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14202493 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14202537 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14202511 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10461285 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461297 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461292 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461301 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461299 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461280 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461281 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461287 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461289 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461303 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461290 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461294 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461293 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461296 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461282 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461286 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461278 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461279 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461302 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461288 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461295 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461284 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461283 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461300 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461298 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461291 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1437812_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1415787_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10461277 | - | - |
| Mouse Genome 430 2.0 Array | 1437812_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1415787_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1415787_at | - | - |
| Mouse Genome 430 2.0 Array | 1437812_x_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14376_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14376_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900002209.mm.1 | NM_008060.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900002209.mm.1 | TC1900000165.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1437812_PM_x_at | - | - |
| Mouse Expression Set 430 | 1415787_PM_at | - | - |
| Mouse Expression Set 430 | 1415787_at | - | - |
| Mouse Expression Set 430 | 1437812_x_at | - | - |
| Murine Genome U74v2 Set | 97516_PM_at | - | - |
| Murine Genome U74v2 Set | 97516_at | - | - |
| Mouse Genome 430A 2.0 Array | 1437812_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1415787_at | - | - |
| Mouse Genome 430A 2.0 Array | 1437812_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1415787_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1437812_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1415787_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1415787_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1437812_PM_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 460733 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 104060270 | - | - |
| MouseWG-6 v2 Expression BeadChip | 1980400 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 104060300 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5720717 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 104060270 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1980400 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005720717 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001980400 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1237089 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1225535 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1251640 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1225535 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1231453 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1237089 | - | - |
| mouse-6 v1.1 (Illumina) | scl014376.17_49-S | - | - |
| mouse-6 v1.1 (Illumina) | scl53398.18.1_7-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5720717 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1980400 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 104060270 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 104060300 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl014376.17_49-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl53398.18.1_7-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5720717 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 104060300 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 104060270 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1980400 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1237089 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2940440 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1225535 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1231453 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1225535 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1251640 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2940440 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1237089 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl53398.18.1_7-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl014376.17_49-S | - | - |

Ganab
Genomic Map