Assay Details
Gene Symbol: Gapdhs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290631.1 | NP_001277560.1 | 8 - 9 | 1017 | 69 | |
NM_008085.2 | NP_032111.1 | 8 - 9 | 1035 | 69 | ||
GenBank mRNA | AK131805.1 | - | 4 - 5 | 378 | 69 | |
M60978.1 | - | 8 - 9 | 1017 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
