Assay Details
Gene Symbol: Mgp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008597.3 | NP_032623.1 | 1 - 2 | 132 | 105 | |
GenBank mRNA | AK018719.1 | - | 1 - 2 | 133 | 105 | |
AK159591.1 | - | 1 - 2 | 136 | 105 | ||
BC036991.1 | - | 1 - 2 | 128 | 4990627 | 105 | |
BC069922.1 | - | 1 - 2 | 122 | 4984262 | 105 | |
BC079478.1 | - | 1 - 2 | 123 | 4950210 | 105 | |
BY702829.1 | - | 1 - 2 | 133 | 105 | ||
D00613.1 | - | 1 - 2 | 75 | 105 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK018719.1,AK159591.1,BC036991.1,BC069922.1,BC079478.1,D00613.1,NM_008597.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P426270 | AK159591.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P426270 | BC018338.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P426270 | BC036991.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P426270 | BC069922.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P426270 | BC079478.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P426270 | AK159591.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P426270 | BC018338.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P426270 | BC036991.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P426270 | BC069922.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P426270 | BC079478.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 910537 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 907383 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 355533 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P170570 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P426270 | AK159591.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P426270 | BC018338.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P426270 | BC036991.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P426270 | BC069922.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P426270 | BC079478.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P426270 | AK159591.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P426270 | BC018338.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P426270 | BC036991.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P426270 | BC069922.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P426270 | BC079478.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P426270 | AK159591.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P426270 | BC018338.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P426270 | BC036991.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P426270 | BC069922.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P426270 | BC079478.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P426270 | AK159591.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P426270 | BC018338.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P426270 | BC036991.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P426270 | BC069922.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P426270 | BC079478.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874947 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874963 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24875002 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874967 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874980 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874989 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24875001 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874958 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874997 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874974 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874943 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874981 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874952 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874951 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874985 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874949 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874978 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874961 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874972 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874941 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874966 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874992 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874959 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874996 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874948 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874983 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874940 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874953 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24875003 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874976 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874994 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874945 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874956 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874977 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874999 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874970 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874968 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874954 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P170570 | - | - |
Whole Mouse Genome G4122A | A_51_P426270 | AK159591.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P426270 | BC018338.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P426270 | BC036991.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P426270 | BC069922.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P426270 | BC079478.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P426270 | AK159591.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P426270 | BC018338.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P426270 | BC036991.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P426270 | BC069922.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P426270 | BC079478.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121119 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P170570 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05148666 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05148652 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05148654 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05148659 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05148647 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05148642 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05148636 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874873 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874888 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875004 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874880 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875022 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874943 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874917 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874959 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874897 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874890 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874994 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874932 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875016 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875007 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874937 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874872 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874886 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874961 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874989 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874967 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874990 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874992 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874976 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874999 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874911 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874963 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874912 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875018 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874981 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875012 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874978 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874895 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875001 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875009 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874876 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874980 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874930 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875014 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874947 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874934 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874972 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874945 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875019 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874910 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874984 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874970 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874997 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874875 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874909 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874878 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874953 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874923 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874974 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874958 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874949 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874952 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875003 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874940 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875005 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874956 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874935 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24875002 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874920 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874977 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874907 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874922 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874915 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874925 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874889 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874883 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874922 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24875009 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24875003 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874925 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874911 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874905 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05148651 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874981 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24875016 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05832559 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874989 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874952 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874907 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874967 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24875007 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24875006 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24875018 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874997 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874994 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874916 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874943 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874926 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874947 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24875014 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874978 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874977 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874992 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874984 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874923 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874940 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874968 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874928 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874974 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874970 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874990 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24875019 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874935 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05277048 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874999 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24875005 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874919 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874956 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05691990 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874932 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05650458 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874920 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05290684 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05768368 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874961 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874953 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874972 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874980 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874945 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874934 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24875004 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05768378 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24875002 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874914 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874959 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874930 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05277047 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874915 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874949 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874963 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874910 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10548881 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10548883 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10548882 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10548880 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1448416_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10548879 | - | - |
Mouse Genome 430 2.0 Array | 1448416_at | - | - |
Mouse Genome 430 2.0 Array | 1448416_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 17313_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 17313_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0600026168.mm.1 | TC0600003335.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600026168.mm.1 | NM_008597.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600026170.mm.1 | TC0600003335.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600026170.mm.1 | NM_008597.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600026169.mm.1 | TC0600003335.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600026169.mm.1 | NM_008597.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600026171.mm.1 | TC0600003335.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600026171.mm.1 | NM_008597.4 | Refseq |
Mouse Expression Set 430 | 1448416_PM_at | - | - |
Mouse Expression Set 430 | 1448416_at | - | - |
Murine Genome U74v2 Set | 93866_PM_s_at | - | - |
Murine Genome U74v2 Set | 93866_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1448416_at | - | - |
Mouse Genome 430A 2.0 Array | 1448416_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1448416_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1448416_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 610341 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 6900736 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006900736 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2666018 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2666018 | - | - |
mouse-6 v1.1 (Illumina) | scl017313.2_5-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6900736 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl017313.2_5-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6900736 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2666018 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2666018 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl017313.2_5-S | - | - |
