Assay Details
| Gene Symbol: Mpz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008623.4 | NP_032649.2 | 7 - 8 | 814 | 97 | |
| GenBank mRNA | AK089180.1 | - | 7 - 8 | 818 | 97 | |
| AK157960.1 | - | 5 - 6 | 711 | 97 | ||
| BC139139.1 | - | 5 - 6 | 679 | 8862161 | 97 | |
| BC141226.1 | - | 5 - 6 | 679 | 9055838 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2001223 | BC139139.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2001223 | BC141226.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2001223 | NM_008623.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2001223 | AK157960.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 728498 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 182182 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 38342 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 435854 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 257247 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 818199 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 102596 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 27852 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P135358 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2001223 | BC139139.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2001223 | BC141226.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2001223 | NM_008623.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2001223 | AK157960.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P321235 | BC139139.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P321235 | BC141226.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P321235 | NM_008623.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P321235 | AK089180.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P321235 | AK157960.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826340 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826348 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826346 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826349 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826344 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826353 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826351 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826336 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826363 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826360 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826358 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826354 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826355 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826335 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826356 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826368 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826342 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826366 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826337 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P135358 | - | - |
| Whole Mouse Genome G4122A | A_51_P321235 | BC139139.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P321235 | BC141226.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P321235 | NM_008623.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P321235 | AK089180.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P321235 | AK157960.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P126715 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124169 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P135358 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00453038 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00453039 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00453040 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00373229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826405 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826392 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826416 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826354 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826346 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826412 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00775923 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00675893 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00891536 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01002780 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826356 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00373229 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826337 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826342 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826405 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826370 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826382 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00775919 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826410 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826373 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826378 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826334 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826340 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826350 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00018980 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826349 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826363 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826383 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826385 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00018982 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00775922 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826366 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826419 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826368 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00891537 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826344 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826397 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826372 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826391 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826360 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826414 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00060026 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826353 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00597652 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826417 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826336 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826381 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826389 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826348 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826415 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00597650 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826380 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826384 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00891533 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826388 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826408 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10351531 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351529 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351530 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351526 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351532 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351527 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351528 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423253_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10351525 | - | - |
| Mouse Genome 430 2.0 Array | 1423253_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1423253_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 17528_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 17528_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100012920.mm.1 | NM_008623.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100012920.mm.1 | TC0100001578.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100012922.mm.1 | NM_008623.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100012922.mm.1 | TC0100001578.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100012918.mm.1 | NM_008623.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100012918.mm.1 | TC0100001578.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100012921.mm.1 | NM_008623.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100012921.mm.1 | TC0100001578.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1423253_PM_at | - | - |
| Mouse Expression Set 430 | 1423253_at | - | - |
| Murine Genome U74v2 Set | 93715_PM_at | - | - |
| Murine Genome U74v2 Set | 93715_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423253_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423253_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423253_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423253_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 558664 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1990333 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1990333 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001990333 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1231021 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1231021 | - | - |
| mouse-6 v1.1 (Illumina) | scl017528.3_10-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1990333 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl017528.3_10-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1990333 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2817329 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1231021 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2817329 | - | - |

Mpz
Genomic Map