Assay Details
Gene Symbol: Mt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008631.2 | NP_032657.1 | 2 - 3 | 134 | 80 | |
GenBank mRNA | AK131894.1 | - | 2 - 3 | 134 | 80 | |
BC099850.1 | - | 2 - 3 | 93 | 80 | ||
BC099851.1 | - | 2 - 3 | 94 | 80 | ||
BC099852.1 | - | 2 - 3 | 94 | 80 | ||
BC099853.1 | - | 2 - 3 | 93 | 80 | ||
BC111805.1 | - | 2 - 3 | 87 | 80 | ||
BC141870.1 | - | 2 - 3 | 95 | 40038882 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P424272 | BC141870.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P424272 | BC099852.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P424272 | BC099853.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P424272 | BC111805.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P424272 | NM_008631.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P424272 | BC141870.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P424272 | BC099852.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P424272 | BC099853.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P424272 | BC111805.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P424272 | NM_008631.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 961164 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 186289 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P138343 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P424272 | BC141870.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P424272 | BC099852.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P424272 | BC099853.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P424272 | BC111805.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P424272 | NM_008631.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P424272 | BC141870.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P424272 | BC099852.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P424272 | BC099853.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P424272 | BC111805.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P424272 | NM_008631.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075679 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075681 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075695 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075662 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075657 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075665 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075671 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075693 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075691 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075677 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075689 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075686 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075674 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075675 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075670 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075680 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075669 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075687 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075659 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075673 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075696 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075694 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075688 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075667 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075692 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075684 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P138343 | - | - |
Whole Mouse Genome G4122A | A_51_P424272 | BC141870.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P424272 | BC099852.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P424272 | BC099853.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P424272 | BC111805.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P424272 | NM_008631.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P424272 | BC141870.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P424272 | BC099852.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P424272 | BC099853.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P424272 | BC111805.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P424272 | NM_008631.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P424275 | BC141870.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P424275 | BC099852.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P424275 | BC099853.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P424275 | BC111805.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P424275 | NM_008631.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134996 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P138343 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06945278 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06962495 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06962489 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06962490 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075671 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075711 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075633 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075651 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075638 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075647 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075686 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075687 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075645 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075688 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075736 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075673 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075629 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075640 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075691 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075744 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075737 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075657 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075677 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075708 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075689 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075636 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075702 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075679 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075741 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075734 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075659 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075639 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075732 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075693 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075661 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075674 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075631 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075692 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075699 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075680 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075665 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075643 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075669 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075662 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075701 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075707 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075681 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075652 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075684 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075705 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075667 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075695 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26075680 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06962495 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26075645 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26075659 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26075674 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26075707 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26075631 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06808293 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26075677 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26075639 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26075701 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07304811 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07246482 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26075702 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06865914 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26075633 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26075669 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26075670 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26075675 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07369557 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07160540 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06962489 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26075649 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06853465 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06766767 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26075693 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26075648 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07072796 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26075686 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07225643 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07441894 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26075642 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26075640 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06808287 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26075687 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26075652 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26075673 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07086042 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26075667 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26075656 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26075684 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07441895 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26075627 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26075671 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07058162 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07304805 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06766683 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26075689 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26075696 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10574015 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10574017 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10574016 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10574014 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1450645_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10574013 | - | - |
Mouse Genome 430 2.0 Array | 1450645_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1450645_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 17752_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 17752_PM_at | - | - |
Mouse Expression Set 430 | 1450645_PM_at | - | - |
Mouse Expression Set 430 | 1450645_at | - | - |
Murine Genome U74v2 Set | 102989_at | - | - |
Murine Genome U74v2 Set | 102989_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1450645_at | - | - |
Mouse Genome 430A 2.0 Array | 1450645_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1450645_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1450645_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 454125 | - | - |
ABI Mouse Genome Survey Microarray | 707078 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 4780338 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004780338 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2740391 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2740391 | - | - |
mouse-6 v1.1 (Illumina) | scl017752.3_0-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4780338 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl017752.3_0-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4780338 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2740391 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2740391 | - | - |
