Assay Details
| Gene Symbol: Rbpjl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009036.1 | NP_033062.1 | 7 - 8 | 855 | 131 | |
| GenBank mRNA | AK052449.1 | - | 5 - 6 | 3966 | 131 | |
| AK131814.1 | - | 7 - 8 | 836 | 131 | ||
| AK131832.1 | - | 7 - 8 | 792 | 131 | ||
| BC152311.1 | - | 7 - 8 | 849 | 40090714 | 131 | |
| Y10926.1 | - | 7 - 8 | 855 | 131 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rbpjl
Genomic Map