Assay Details
| Gene Symbol: Reg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009042.1 | NP_033068.1 | 4 - 5 | 396 | 99 | |
| GenBank mRNA | AK075798.1 | - | 2 - 3 | 209 | 99 | |
| AK133506.1 | - | 4 - 5 | 400 | 99 | ||
| AK160452.1 | - | 4 - 5 | 397 | 99 | ||
| BC028761.1 | - | 4 - 5 | 392 | 1263580 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P329811 | NM_009042.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P329811 | AK075798.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P329811 | AK133506.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P329811 | BC028761.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P329811 | NM_009042.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P329811 | AK075798.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P329811 | AK133506.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P329811 | BC028761.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 401096 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 173933 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 589100 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 51624 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P147383 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P329811 | NM_009042.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P329811 | AK075798.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P329811 | AK133506.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P329811 | BC028761.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P329811 | NM_009042.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P329811 | AK075798.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P329811 | AK133506.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P329811 | BC028761.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P329811 | NM_009042.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P329811 | AK075798.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P329811 | AK133506.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P329811 | BC028761.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P329811 | NM_009042.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P329811 | AK075798.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P329811 | AK133506.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P329811 | BC028761.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24570041 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24570051 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24570057 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24570067 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24570068 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24570036 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24570035 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24570039 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24570045 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24570046 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24570029 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24570052 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24570034 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24570032 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24570062 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24570061 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24570053 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24570064 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24570037 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24570063 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24570055 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24570048 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24570050 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24570043 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24570066 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24570042 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24570054 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24570056 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24570059 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P147383 | - | - |
| Whole Mouse Genome G4122A | A_51_P329811 | NM_009042.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P329811 | AK075798.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P329811 | AK133506.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P329811 | BC028761.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P329811 | NM_009042.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P329811 | AK075798.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P329811 | AK133506.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P329811 | BC028761.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P104605 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P147383 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05354091 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05354076 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05354070 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05354072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24570023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24570055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24570067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24570051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24570057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24570034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24570076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24570075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24570059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24570062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24570046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24570053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24570018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24570085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24570061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24570072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24570032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24570050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24570048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24570064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24570082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24570041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24570027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24570073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24570056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24570052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24570039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24570083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24570020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24570043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24570022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24570030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24570036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24570069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24570037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24570063 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24570034 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24570023 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05352934 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24570020 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05496147 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24570039 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24570076 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24570069 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24570043 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24570046 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24570057 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05354080 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24570064 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05354082 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24570027 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24570078 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24570056 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24570085 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24570018 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05673296 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24570072 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24570063 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05352937 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24570032 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24570053 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24570079 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24570077 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24570051 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24570066 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24570073 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24570062 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24570022 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05354089 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24570050 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24570041 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24570030 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24570059 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05354087 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05354095 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05270871 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05088980 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24570036 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10539205 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539206 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539201 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539202 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539204 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539203 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1415905_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10539200 | - | - |
| Mouse Genome 430 2.0 Array | 1415905_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1415905_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19692_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19692_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600006687.mm.1 | TC0600000878.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600006687.mm.1 | NM_009042.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600006687.mm.1 | NM_009042.1 | Refseq |
| Mouse Expression Set 430 | 1415905_at | - | - |
| Mouse Expression Set 430 | 1415905_PM_at | - | - |
| Murine Genome U74v2 Set | 162312_f_at | - | - |
| Murine Genome U74v2 Set | 162312_PM_f_at | - | - |
| Murine Genome U74v2 Set | 160213_PM_at | - | - |
| Murine Genome U74v2 Set | 160213_at | - | - |
| Mouse Genome 430A 2.0 Array | 1415905_at | - | - |
| Mouse Genome 430A 2.0 Array | 1415905_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1415905_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1415905_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 688222 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3610452 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3610452 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003610452 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2592415 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2592415 | - | - |
| mouse-6 v1.1 (Illumina) | scl019692.4_40-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3610452 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl019692.4_40-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3610452 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2592415 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2592415 | - | - |
Citations (1)
Interleukin-18, together with interleukin-12, induces severe acute pancreatitis in obese but not in nonobese leptin-deficient mice.
- Authors
- Sennello JA, Fayad R, Pini M, Gove ME, Ponemone V, Cabay RJ, Siegmund B, Dinarello CA, Fantuzzi G
- Journal
- Proc Natl Acad Sci U S A - Page No 8085 (Tue Jun 10 00:00:00 EDT 2008)
- Volume (Year)
- 105 (2008)
- Product Usage
- -

Reg1
Genomic Map