Assay Details
Gene Symbol: Slc50a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009057.3 | NP_033083.2 | 2 - 3 | 262 | 73 | |
GenBank mRNA | AK014305.1 | - | 2 - 3 | 215 | 73 | |
AK170162.1 | - | 2 - 3 | 213 | 73 | ||
BC014292.1 | - | 2 - 3 | 246 | 4190124 | 73 | |
BY333933.1 | - | 2 - 3 | 133 | 73 | ||
X96618.1 | - | 2 - 3 | 253 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
