Assay Details
Gene Symbol: Slc50a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009057.3 | NP_033083.2 | 4 - 5 | 550 | 75 | |
GenBank mRNA | AK014305.1 | - | 4 - 5 | 503 | 75 | |
AK045924.1 | - | 2 - 3 | 863 | 75 | ||
AK079716.1 | - | 3 - 4 | 615 | 75 | ||
AK158862.1 | - | 3 - 4 | 575 | 75 | ||
AK170162.1 | - | 4 - 5 | 501 | 75 | ||
BC014292.1 | - | 4 - 5 | 534 | 4190124 | 75 | |
X96618.1 | - | 4 - 5 | 541 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
