Assay Details
| Gene Symbol: Polr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009086.2 | NP_033112.2 | 7 - 8 | 1241 | 94 | |
| GenBank mRNA | AK083732.1 | - | 7 - 8 | 1223 | 94 | |
| AK143501.1 | - | 7 - 8 | 1178 | 94 | ||
| AK144616.1 | - | 7 - 8 | 1228 | 94 | ||
| AK156712.1 | - | 7 - 8 | 1232 | 94 | ||
| BC060656.1 | - | 7 - 8 | 1171 | 30355130 | 94 | |
| U58280.1 | - | 7 - 8 | 1205 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Polr1b
Genomic Map