Assay Details
| Gene Symbol: Frrs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113478.1 | NP_001106950.1 | 8 - 9 | 1036 | 102 | |
| NM_009146.3 | NP_033172.2 | 8 - 9 | 998 | 102 | ||
| GenBank mRNA | AK075573.1 | - | 8 - 9 | 990 | 102 | |
| AK144933.1 | - | 8 - 9 | 990 | 102 | ||
| AK162693.1 | - | 8 - 9 | 991 | 102 | ||
| AK167289.1 | - | 8 - 9 | 1026 | 102 | ||
| BC027770.1 | - | 7 - 8 | 996 | 5344922 | 102 | |
| D50464.1 | - | 8 - 9 | 986 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Frrs1
Genomic Map