Assay Details
| Gene Symbol: Shc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009167.3 | NP_033193.2 | 4 - 5 | 367 | 78 | |
| GenBank mRNA | BC103611.1 | - | 4 - 5 | 367 | 40046304 | 78 |
| BC103612.1 | - | 4 - 5 | 367 | 40046306 | 78 | |
| BC105644.1 | - | 4 - 5 | 367 | 40046301 | 78 | |
| BC105645.1 | - | 4 - 5 | 367 | 40046303 | 78 | |
| U46854.1 | - | 4 - 5 | 367 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Shc3
Genomic Map