Assay Details
| Gene Symbol: Soat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009230.3 | NP_033256.2 | 9 - 10 | 1116 | 72 | |
| GenBank mRNA | AK052299.1 | - | 6 - 7 | 673 | 72 | |
| AK142486.1 | - | 9 - 10 | 1033 | 72 | ||
| AK159393.1 | - | 9 - 10 | 1046 | 72 | ||
| AK159529.1 | - | 9 - 10 | 1046 | 72 | ||
| AK160017.1 | - | 9 - 10 | 1046 | 72 | ||
| BC083092.1 | - | 8 - 9 | 930 | 30446741 | 72 | |
| DQ903181.1 | - | 9 - 10 | 1033 | 72 | ||
| S81092.1 | - | 9 - 10 | 1050 | 72 | ||
More Information
Set Membership: |
|

Soat1
Genomic Map