Assay Details
| Gene Symbol: Car6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009802.2 | NP_033932.2 | 6 - 7 | 748 | 95 | |
| GenBank mRNA | AF079834.1 | - | 5 - 6 | 838 | 95 | |
| AF079835.1 | - | 6 - 7 | 775 | 95 | ||
| AK132599.1 | - | 6 - 7 | 749 | 95 | ||
| AK161216.1 | - | 6 - 7 | 744 | 95 | ||
| BC006726.1 | - | 6 - 7 | 835 | 95 | ||
| BC018582.1 | - | 6 - 7 | 725 | 95 | ||
| BC036270.1 | - | 6 - 7 | 731 | 95 | ||
| BC039657.1 | - | 6 - 7 | 733 | 95 | ||
| BC046495.1 | - | 5 - 6 | 524 | 95 | ||
| BC049973.1 | - | 6 - 7 | 746 | 95 | ||
| BC055437.1 | - | 6 - 7 | 734 | 4190566 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2004370 | BC049973.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2004370 | BC055437.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2004370 | BC018582.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2004370 | BC046495.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2004370 | AK132599.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 181832 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 952234 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 162629 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 885946 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 575156 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 262417 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 973173 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 274659 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 740883 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 788975 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 16718 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 136890 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P124702 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P175420 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2004370 | BC049973.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2004370 | BC055437.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2004370 | BC018582.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2004370 | BC046495.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2004370 | AK132599.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P310511 | BC049973.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P310511 | BC055437.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P310511 | BC018582.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P310511 | BC046495.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P310511 | AF079835.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23404821 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23404831 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23404846 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23404858 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23404828 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23404835 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23404851 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23404823 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23404853 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23404833 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23404850 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23404836 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23404854 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23404861 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23404818 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23404830 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23404847 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23404829 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23404838 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23404842 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23404837 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23404856 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23404841 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23404825 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23404845 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23404839 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23404826 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P175420 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P124702 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05449141 | - | - |
| Whole Mouse Genome G4122A | A_52_P310511 | BC049973.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P310511 | BC055437.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P310511 | BC018582.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P310511 | BC046495.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P310511 | AF079835.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109475 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P124702 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P175420 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03982295 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03982315 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03982307 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03982296 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03982300 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03982305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404760 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404810 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03472881 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03730768 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03996946 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404791 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03982299 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04081431 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404795 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404803 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03982301 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03703042 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04012892 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404804 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03982295 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404858 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404801 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03375323 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404828 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404846 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404842 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404857 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404780 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404796 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03324048 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404872 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404845 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03280836 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04072393 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03375325 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03982296 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03982313 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404837 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03996949 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03288838 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404783 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404874 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04080138 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404826 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404843 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404775 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404853 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404831 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404838 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03324044 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404800 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03222098 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03444268 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404779 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404833 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04069416 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404785 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03444266 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404856 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404789 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404869 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404847 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404818 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03820488 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404835 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03324045 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404798 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404873 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404854 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03444269 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404776 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03454632 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04081430 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404864 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10518752 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10518759 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10518758 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10518754 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10518757 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10518760 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10518753 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10518756 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10518755 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1421001_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10518751 | - | - |
| Mouse Genome 430 2.0 Array | 1421001_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1421001_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12353_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12353_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400033766.mm.1 | TC0400004062.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400033766.mm.1 | NM_009802.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400033766.mm.1 | NM_009802.2 | Refseq |
| Mouse Expression Set 430 | 1421001_PM_a_at | - | - |
| Mouse Expression Set 430 | 1421001_a_at | - | - |
| Murine Genome U74v2 Set | 160647_at | - | - |
| Murine Genome U74v2 Set | 160647_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421001_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421001_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421001_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421001_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 922984 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 102340487 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 102340487 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5290017 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005290017 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1257323 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2489268 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1257323 | - | - |
| mouse-6 v1.1 (Illumina) | scl012353.1_324-S | - | - |
| mouse-6 v1.1 (Illumina) | scl23485.1.124_12-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5290017 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 102340487 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl012353.1_324-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5290017 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102340487 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1257323 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1257323 | - | - |

Car6
Genomic Map