Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100001886.mm.1 |
NM_009837.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100001886.mm.1 |
NM_009837.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100001886.mm.1 |
TC1100000234.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100001891.mm.1 |
NM_009837.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100001891.mm.1 |
NM_009837.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100001891.mm.1 |
TC1100000234.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100001888.mm.1 |
NM_009837.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100001888.mm.1 |
NM_009837.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100001888.mm.1 |
TC1100000234.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100001892.mm.1 |
NM_009837.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100001892.mm.1 |
NM_009837.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100001892.mm.1 |
TC1100000234.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100001895.mm.1 |
NM_009837.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100001895.mm.1 |
NM_009837.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100001895.mm.1 |
TC1100000234.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100001893.mm.1 |
NM_009837.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100001893.mm.1 |
NM_009837.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100001893.mm.1 |
TC1100000234.mm.1 |
MTA TC |