Assay Details
| Gene Symbol: Cct8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009840.3 | NP_033970.3 | 10 - 11 | 1178 | 129 | |
| GenBank mRNA | AK010882.1 | - | 10 - 11 | 1179 | 129 | |
| AK014005.1 | - | 3 - 4 | 244 | 129 | ||
| AK075711.1 | - | 10 - 11 | 1159 | 129 | ||
| AK075880.1 | - | 10 - 11 | 1167 | 129 | ||
| AK145661.1 | - | 10 - 11 | 1162 | 129 | ||
| AK145754.1 | - | 10 - 11 | 1161 | 129 | ||
| AK145918.1 | - | 10 - 11 | 1180 | 129 | ||
| AK147038.1 | - | 10 - 11 | 1181 | 129 | ||
| AK159969.1 | - | 10 - 11 | 128 | |||
| AK168465.1 | - | 10 - 11 | 1151 | 129 | ||
| AK172867.1 | - | 10 - 11 | 1149 | 129 | ||
| AK218945.1 | - | 3 - 4 | 205 | 129 | ||
| BC009007.1 | - | 10 - 11 | 1136 | 3582639 | 129 | |
| Z37164.1 | - | 10 - 11 | 1182 | 129 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cct8
Genomic Map