Assay Details
Gene Symbol: Cdh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009864.2 | NP_033994.1 | 14 - 15 | 2427 | 87 | |
GenBank mRNA | AK166267.1 | - | 14 - 15 | 2429 | 87 | |
BC098501.1 | - | 14 - 15 | 2416 | 30023851 | 87 | |
X06115.1 | - | 14 - 15 | 2368 | 87 | ||
X06339.1 | - | 11 - 12 | 1781 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
