Assay Details
| Gene Symbol: Cdh4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009867.2 | NP_033997.1 | 4 - 5 | 705 | 109 | |
| GenBank mRNA | BC047991.1 | - | 4 - 5 | 705 | 109 | |
| D14888.1 | - | 4 - 5 | 615 | 109 | ||
| X69966.1 | - | 4 - 5 | 561 | 109 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdh4
Genomic Map