Assay Details
| Gene Symbol: Cdkn2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009878.3 | NP_034008.2 | 1 - 2 | 401 | 59 | |
| GenBank mRNA | AK132906.1 | - | 1 - 2 | 366 | 59 | |
| AK140808.1 | - | 1 - 2 | 78 | 59 | ||
| BC013898.1 | - | 1 - 2 | 230 | 4020283 | 59 | |
| U19597.1 | - | 1 - 2 | 274 | 59 | ||
| U20497.1 | - | 1 - 2 | 168 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cyclin-dependent protein serine/threonine kinase activity
G1/S transition of mitotic cell cycle
DNA synthesis involved in DNA repair
cell cycle
cell cycle arrest
sensory perception of sound
negative regulation of cell proliferation
response to UV
negative regulation of cell growth
response to retinoic acid
response to vitamin D
negative regulation of phosphorylation
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
autophagic cell death
negative regulation of protein serine/threonine kinase activity
negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage
cyclin-dependent protein serine/threonine kinase inhibitor activity
protein kinase binding
regulation of cyclin-dependent protein serine/threonine kinase activity
G1/S transition of mitotic cell cycle
DNA synthesis involved in DNA repair
cell cycle
cell cycle arrest
sensory perception of sound
negative regulation of cell proliferation
response to UV
negative regulation of cell growth
response to retinoic acid
response to vitamin D
negative regulation of phosphorylation
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
autophagic cell death
negative regulation of protein serine/threonine kinase activity
negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage
cyclin-dependent protein serine/threonine kinase inhibitor activity
protein kinase binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26346397 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26346391 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26346395 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26346392 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26346396 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26346398 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26346390 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26346394 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26346393 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2022251 | BC013898.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2022251 | AK140808.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2022251 | AK144737.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2022251 | U20497.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2022251 | NM_009878.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2022251 | BC013898.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2022251 | AK140808.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2022251 | AK144737.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2022251 | U20497.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2022251 | NM_009878.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P122356 | AK140808.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P122356 | AK144737.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P122356 | U20497.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P122356 | NM_009878.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346397 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346401 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346412 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346396 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346378 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346374 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346395 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346398 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346373 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346381 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346388 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346392 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346409 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346376 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346385 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346375 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346383 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346390 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346393 | - | - |
| Whole Mouse Genome G4122A | A_51_P122356 | AK140808.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122356 | AK144737.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122356 | U20497.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P122356 | NM_009878.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108577 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131942 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07471982 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07471966 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07471973 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07471975 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07470103 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07470099 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26346393 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26346390 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26346392 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26346395 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26346394 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26346398 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26346396 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26346397 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26346391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26897492 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346397 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26897499 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07658155 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07470099 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07664194 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26897471 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346378 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07975996 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07552772 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26897476 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07474022 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346383 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26897518 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26897492 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07975991 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26897520 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346381 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346413 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26897513 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26897495 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07664192 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346388 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26897494 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346395 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07552773 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26897497 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26897501 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07648934 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346373 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26897483 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26897514 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07471975 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346390 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26897498 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08067128 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07749282 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346385 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346411 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346375 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26897500 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08067129 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26897493 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346401 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346392 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26897484 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08095738 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26897486 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07474025 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26897489 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26897522 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26897504 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10591521 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591518 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591520 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591519 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10591517 | - | - |
| Mouse Genome 430 2.0 Array | 1416253_at | - | - |
| Mouse Genome 430 2.0 Array | 1416253_PM_at | - | - |
| Affymetrix Murine Genome 6500A Array | Msa15380_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900015716.mm.1 | NM_009878.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900015716.mm.1 | NM_009878.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900015716.mm.1 | TC0900001867.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900015718.mm.1 | NM_009878.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900015718.mm.1 | NM_009878.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900015718.mm.1 | TC0900001867.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900015719.mm.1 | NM_009878.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900015719.mm.1 | NM_009878.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900015719.mm.1 | TC0900001867.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1416253_at | - | - |
| Mouse Expression Set 430 | 1416253_PM_at | - | - |
| Murine Genome U74v2 Set | 94521_at | - | - |
| Murine Genome U74v2 Set | 94521_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416253_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416253_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 827619 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6040035 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6040035 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 102260154 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006040035 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1225214 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1225214 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2443959 | - | - |
| mouse-6 v1.1 (Illumina) | scl36146.3_205-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0003491.1_29-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 102260154 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6040035 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl36146.3_205-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102260154 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6040035 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1225214 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1225214 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl36146.3_205-S | - | - |
Citations (1)
Expression of p16Ink4a compensates for p18Ink4c loss in cyclin-dependent kinase 4/6-dependent tumors and tissues.
- Authors
- Ramsey MR, Krishnamurthy J, Pei XH, Torrice C, Lin W, Carrasco DR, Ligon KL, Xiong Y, Sharpless NE
- Journal
- Cancer Res - Page No 4732 (Tue May 15 00:00:00 EDT 2007)
- Volume (Year)
- 67 (2007)
- Product Usage
- -

Cdkn2d
Genomic Map