Assay Details
| Gene Symbol: Cel | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009885.1 | NP_034015.1 | 7 - 8 | 889 | 69 | |
| GenBank mRNA | AK131818.1 | - | 7 - 8 | 907 | 69 | |
| AK131824.1 | - | 7 - 8 | 907 | 69 | ||
| BC006872.1 | - | 7 - 8 | 901 | 2646703 | 69 | |
| U33169.1 | - | 7 - 8 | 889 | 69 | ||
| U37386.1 | - | 7 - 8 | 893 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cel
Genomic Map