Assay Details
Gene Symbol: Cnn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009922.4 | NP_034052.3 | 1 - 2 | 167 | 59 | |
GenBank mRNA | AK035548.1 | - | 1 - 2 | 140 | 59 | |
BC138863.1 | - | 1 - 2 | 99 | 8861885 | 59 | |
BC138864.1 | - | 1 - 2 | 99 | 8861886 | 59 | |
BC145446.1 | - | 1 - 2 | 99 | 59 | ||
BY326336.1 | - | 1 - 2 | 205 | 59 | ||
Z19542.1 | - | 1 - 2 | 161 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P350817 | Z19542.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P350817 | NM_009922.4 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P350817 | AK035548.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P350817 | Z19542.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P350817 | NM_009922.4 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P350817 | AK035548.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 503023 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 130421 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 780207 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 869258 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 963785 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 910606 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 900155 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 9571 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P159198 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P107877 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P350817 | Z19542.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P350817 | NM_009922.4 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P350817 | AK035548.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P350817 | Z19542.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P350817 | NM_009922.4 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P350817 | AK035548.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P350817 | Z19542.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P350817 | NM_009922.4 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P350817 | AK035548.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P350817 | Z19542.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P350817 | NM_009922.4 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P350817 | AK035548.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26350427 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26350449 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26350438 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26350453 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26350424 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26350417 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26350421 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26350445 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26350422 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26350433 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26350441 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26350447 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26350446 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26350452 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26350426 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26350455 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26350419 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26350450 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26350423 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26350443 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26350436 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26350440 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26350448 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26350430 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P107877 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P159198 | - | - |
Whole Mouse Genome G4122A | A_51_P350817 | Z19542.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P350817 | NM_009922.4 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P350817 | AK035548.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P350817 | Z19542.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P350817 | NM_009922.4 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P350817 | AK035548.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134050 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P107877 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P159198 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07907835 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07907838 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350524 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350462 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350535 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350443 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350523 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350445 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350466 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350441 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350419 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350414 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350422 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350476 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350494 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350473 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350433 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350426 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350450 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350484 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350530 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350486 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350453 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350481 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350508 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350528 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350514 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350479 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350467 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350496 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350428 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350493 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350440 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350538 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350513 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350489 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350437 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350413 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350403 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350507 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350483 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350500 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350510 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350512 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350421 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350490 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350475 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350404 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350471 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350469 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350516 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350533 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350460 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350518 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350446 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350452 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350448 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350531 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350437 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350419 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350486 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350430 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07807343 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350413 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350471 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350464 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07708497 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350453 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350441 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07462334 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350500 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350502 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350428 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350450 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350456 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350422 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350483 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350443 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350462 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350423 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350474 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08085474 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350479 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350493 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350476 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350417 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350489 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350466 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350446 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350457 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07907840 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350452 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350494 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350490 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350475 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350497 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350431 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350414 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350460 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350458 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350445 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350467 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350433 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07662199 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350463 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350484 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07855146 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350469 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350473 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07983827 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350481 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07985511 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07983824 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350416 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08081430 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10583814 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10583815 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10583811 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10583810 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10583813 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10583812 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10583816 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1417917_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10583809 | - | - |
Mouse Genome 430 2.0 Array | 1417917_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1417917_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12797_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12797_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0900001627.mm.1 | NM_009922.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900001627.mm.1 | NM_009922.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900001627.mm.1 | TC0900000212.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0900001626.mm.1 | NM_009922.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900001626.mm.1 | NM_009922.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900001626.mm.1 | TC0900000212.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0900001623.mm.1 | NM_009922.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900001623.mm.1 | NM_009922.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900001623.mm.1 | TC0900000212.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0900001622.mm.1 | NM_009922.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900001622.mm.1 | NM_009922.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900001622.mm.1 | TC0900000212.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0900001625.mm.1 | NM_009922.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900001625.mm.1 | NM_009922.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900001625.mm.1 | TC0900000212.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0900001628.mm.1 | NM_009922.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900001628.mm.1 | NM_009922.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900001628.mm.1 | TC0900000212.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0900001629.mm.1 | NM_009922.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900001629.mm.1 | NM_009922.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900001629.mm.1 | TC0900000212.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0900001632.mm.1 | NM_009922.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900001632.mm.1 | NM_009922.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900001632.mm.1 | TC0900000212.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0900001631.mm.1 | NM_009922.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900001631.mm.1 | NM_009922.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900001631.mm.1 | TC0900000212.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0900001624.mm.1 | NM_009922.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900001624.mm.1 | NM_009922.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900001624.mm.1 | TC0900000212.mm.1 | MTA TC |
Mouse Expression Set 430 | 1417917_at | - | - |
Mouse Expression Set 430 | 1417917_PM_at | - | - |
Murine Genome U74v2 Set | 99942_s_at | - | - |
Murine Genome U74v2 Set | 99942_PM_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1417917_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1417917_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1417917_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1417917_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 570874 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 510707 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0000510707 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1221148 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1221148 | - | - |
mouse-6 v1.1 (Illumina) | scl012797.7_132-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 510707 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl012797.7_132-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 510707 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1221148 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1221148 | - | - |
