Assay Details
| Gene Symbol: Fkbp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163481.1 | NP_001156953.1 | 1 - 2 | 333 | 87 | |
| NM_010221.2 | NP_034351.2 | 1 - 2 | 333 | 87 | ||
| GenBank mRNA | AK019446.2 | - | 1 - 2 | 271 | 87 | |
| AK144185.1 | - | 1 - 2 | 316 | 87 | ||
| BC029546.1 | - | 1 - 2 | 280 | 5059670 | 87 | |
| L07063.1 | - | 1 - 2 | 336 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2089530 | L07063.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 744479 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 344769 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 285422 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 542708 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 22427 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P130373 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2089530 | L07063.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P216965 | BC029546.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P216965 | L07063.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P216965 | AK019446.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P216965 | AK144185.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P216965 | NM_001163481.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28067985 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28068008 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28067984 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28068001 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28067995 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28067991 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28067996 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28067990 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28067988 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28068006 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28067998 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28068009 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28068003 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07463757 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P130373 | - | - |
| Whole Mouse Genome G4122A | A_51_P216965 | BC029546.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P216965 | L07063.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P216965 | AK019446.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P216965 | AK144185.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P216965 | NM_001163481.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108340 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P130373 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09244283 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09244286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28068040 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28067996 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28067985 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28068026 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09244296 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28068031 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28067990 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28068001 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09482959 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28068002 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28067999 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09567429 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28068006 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09244290 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09186557 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28068024 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28068025 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28068021 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28068017 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09244291 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28067998 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28068019 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28068022 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28068012 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28068018 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28068008 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28068028 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28068004 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09244280 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09244284 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09567426 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10381127 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10381124 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10381130 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10381125 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10381123 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10381131 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10381126 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10391131 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10381132 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10381129 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10381128 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1415951_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1449632_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10381122 | - | - |
| Mouse Genome 430 2.0 Array | 1415951_at | - | - |
| Mouse Genome 430 2.0 Array | 1415951_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1449632_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1449632_PM_s_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14230_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14230_PM_at | - | - |
| Affymetrix Murine Genome 6500A Array | Msa17680_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100015024.mm.1 | NM_001163481.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100015024.mm.1 | NM_001163481.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100015024.mm.1 | NM_010221.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100015024.mm.1 | NM_010221.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100015024.mm.1 | TC1100001624.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100015022.mm.1 | NM_001163481.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100015022.mm.1 | NM_001163481.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100015022.mm.1 | NM_010221.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100015022.mm.1 | NM_010221.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100015022.mm.1 | TC1100001624.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100015019.mm.1 | NM_001163481.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100015019.mm.1 | NM_001163481.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100015019.mm.1 | NM_010221.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100015019.mm.1 | NM_010221.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100015019.mm.1 | TC1100001624.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100015021.mm.1 | NM_001163481.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100015021.mm.1 | NM_001163481.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100015021.mm.1 | NM_010221.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100015021.mm.1 | NM_010221.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100015021.mm.1 | TC1100001624.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1415951_PM_at | - | - |
| Mouse Expression Set 430 | 1415951_at | - | - |
| Mouse Expression Set 430 | 1449632_s_at | - | - |
| Mouse Expression Set 430 | 1449632_PM_s_at | - | - |
| Murine Genome U74v2 Set | 162414_f_at | - | - |
| Murine Genome U74v2 Set | 99082_at | - | - |
| Murine Genome U74v2 Set | 99082_PM_at | - | - |
| Murine Genome U74v2 Set | 162414_PM_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1415951_at | - | - |
| Mouse Genome 430A 2.0 Array | 1415951_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449632_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449632_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449632_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449632_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1415951_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1415951_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 423160 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1340706 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001340706 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1215085 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1215085 | - | - |
| mouse-6 v1.1 (Illumina) | scl40910.10_100-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1340706 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl40910.10_100-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1340706 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1215085 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1215085 | - | - |

Fkbp10
Genomic Map