Assay Details
| Gene Symbol: Gamt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010255.3 | NP_034385.1 | 4 - 5 | 522 | 89 | |
| GenBank mRNA | AF015887.1 | - | 4 - 5 | 455 | 89 | |
| AK158087.1 | - | 4 - 5 | 465 | 89 | ||
| BC049233.1 | - | 4 - 5 | 508 | 4195030 | 89 | |
| BE992264.1 | - | 2 - 3 | 89 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Gamt
Genomic Map