Assay Details
| Gene Symbol: Mag | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010758.2 | NP_034888.1 | 1 - 2 | 64 | 72 | |
| GenBank mRNA | AK003029.1 | - | 2 - 3 | 98 | 72 | |
| BC101944.1 | - | 1 - 2 | 37 | 40039200 | 72 | |
| BC103521.1 | - | 1 - 2 | 37 | 40039204 | 72 | |
| BC103522.1 | - | 1 - 2 | 37 | 72 | ||
| BC103523.1 | - | 1 - 2 | 37 | 72 | ||
| M31811.1 | - | 1 - 2 | 64 | 72 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK003029.1,BC101944.1,BC103521.1,BC103522.1,BC103523.1,M31811.1,NM_010758.2 transcripts.
Set Membership: |
|

Mag
Genomic Map