Assay Details
Gene Symbol: Mnt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010813.3 | NP_034943.3 | 2 - 3 | 1068 | 81 | |
GenBank mRNA | AK139743.1 | - | 1 - 2 | 90 | 81 | |
AK147933.1 | - | 2 - 3 | 1052 | 81 | ||
BC054534.1 | - | 2 - 3 | 994 | 5721249 | 81 | |
U77356.1 | - | 2 - 3 | 1068 | 81 | ||
Y07609.1 | - | 2 - 3 | 1014 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
