Assay Details
Gene Symbol: Rlim | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011276.3 | NP_035406.3 | 4 - 5 | 560 | 69 | |
GenBank mRNA | AF069992.1 | - | 4 - 5 | 515 | 69 | |
AK013207.1 | - | 4 - 5 | 532 | 69 | ||
AK029295.1 | - | 4 - 5 | 554 | 69 | ||
BC012960.1 | - | 2 - 3 | 293 | 4193003 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
