Assay Details
| Gene Symbol: Dhrs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172424.1 | NP_001165895.1 | 3 - 4 | 354 | 73 | |
| NM_011303.6 | NP_035433.1 | 2 - 3 | 718 | 73 | ||
| GenBank mRNA | AF061743.1 | - | 2 - 3 | 520 | 73 | |
| AK032958.1 | - | 781 | 73 | |||
| AK151419.1 | - | 2 - 3 | 719 | 73 | ||
| AK198997.1 | - | 1 - 2 | 53 | 73 | ||
| AK200594.1 | - | 1 - 2 | 53 | 73 | ||
| AK205732.1 | - | 1 - 2 | 53 | 73 | ||
| BC010972.1 | - | 2 - 3 | 747 | 4215444 | 73 | |
| BC013540.1 | - | 2 - 3 | 697 | 4241608 | 73 | |
| X95281.1 | - | 2 - 3 | 756 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dhrs3
Genomic Map