Assay Details
| Gene Symbol: Dhrs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172424.1 | NP_001165895.1 | 5 - 6 | 711 | 77 | |
| NM_011303.6 | NP_035433.1 | 4 - 5 | 1075 | 77 | ||
| GenBank mRNA | AK032958.1 | - | 1138 | 77 | ||
| AK087137.1 | - | 4 - 5 | 1151 | 77 | ||
| AK151419.1 | - | 4 - 5 | 1076 | 77 | ||
| BC008980.1 | - | 4 - 5 | 591 | 3587135 | 77 | |
| BC010972.1 | - | 4 - 5 | 1104 | 4215444 | 77 | |
| BC013540.1 | - | 4 - 5 | 1054 | 4241608 | 77 | |
| X95281.1 | - | 4 - 5 | 1113 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dhrs3
Genomic Map