Assay Details
Gene Symbol: Sdf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK014418.1 | - | 2 - 3 | 512 | 71 | |
AK157860.1 | - | 2 - 3 | 377 | 71 | ||
AK163400.1 | - | 2 - 3 | 497 | 71 | ||
AK167113.1 | - | 2 - 3 | 426 | 71 | ||
BC005715.1 | - | 3 - 4 | 646 | 71 | ||
BC068152.1 | - | 2 - 3 | 752 | 30534982 | 71 | |
D50461.1 | - | 2 - 3 | 755 | 71 | ||
U45977.1 | - | 2 - 3 | 660 | 71 | ||
U45978.1 | - | 2 - 3 | 528 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
