Assay Details
| Gene Symbol: St6galnac3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011372.2 | NP_035502.1 | 1 - 2 | 93 | 77 | |
| GenBank mRNA | AK034804.1 | - | 1 - 2 | 95 | 77 | |
| Y11342.1 | - | 1 - 2 | 93 | 77 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK034804.1,NM_011372.2,Y11342.1 transcripts.
Set Membership: |
|

St6galnac3
Genomic Map