Assay Details
Gene Symbol: Slc12a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011390.2 | NP_035520.1 | 1 - 2 | 191 | 93 | |
GenBank mRNA | AF087436.1 | - | 1 - 2 | 193 | 93 | |
AK083598.1 | - | 1 - 2 | 149 | 93 | ||
AK131129.1 | - | 1 - 2 | 145 | 93 | ||
AK143535.1 | - | 1 - 2 | 185 | 93 | ||
AK149750.1 | - | 1 - 2 | 186 | 93 | ||
AK166215.1 | - | 1 - 2 | 185 | 93 | ||
AK171498.1 | - | 1 - 2 | 228 | 93 | ||
BC051558.1 | - | 1 - 2 | 180 | 93 | ||
BC059242.1 | - | 1 - 2 | 185 | 93 | ||
BC060133.1 | - | 1 - 2 | 185 | 93 | ||
BC079677.1 | - | 1 - 2 | 149 | 93 | ||
BC141107.1 | - | 1 - 2 | 151 | 9055719 | 93 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK171498.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
