Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1300005718.mm.1 |
NM_011390.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300005718.mm.1 |
NM_011390.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300005718.mm.1 |
TC1300000887.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300005719.mm.1 |
NM_011390.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300005719.mm.1 |
NM_011390.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300005719.mm.1 |
TC1300000887.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300005720.mm.1 |
NM_011390.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300005720.mm.1 |
NM_011390.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300005720.mm.1 |
TC1300000887.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300005721.mm.1 |
NM_011390.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300005721.mm.1 |
NM_011390.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300005721.mm.1 |
TC1300000887.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300005722.mm.1 |
NM_011390.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300005722.mm.1 |
NM_011390.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300005722.mm.1 |
TC1300000887.mm.1 |
MTA TC |