Assay Details
Gene Symbol: Slc12a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011390.2 | NP_035520.1 | 9 - 10 | 1370 | 78 | |
GenBank mRNA | AF087436.1 | - | 9 - 10 | 1372 | 78 | |
AK083598.1 | - | 6 - 7 | 3641 | 78 | ||
AK131129.1 | - | 9 - 10 | 1324 | 78 | ||
AK143535.1 | - | 9 - 10 | 1364 | 78 | ||
AK149750.1 | - | 9 - 10 | 1365 | 78 | ||
AK166215.1 | - | 9 - 10 | 1364 | 78 | ||
AK169950.1 | - | 9 - 10 | 1360 | 78 | ||
AK171498.1 | - | 9 - 10 | 1407 | 78 | ||
BC059242.1 | - | 9 - 10 | 1362 | 78 | ||
BC060133.1 | - | 9 - 10 | 1362 | 78 | ||
BC079677.1 | - | 9 - 10 | 1328 | 78 | ||
BC141107.1 | - | 9 - 10 | 1330 | 9055719 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
