Assay Details
Gene Symbol: Slc12a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011390.2 | NP_035520.1 | 13 - 14 | 1821 | 73 | |
GenBank mRNA | AF087436.1 | - | 13 - 14 | 1823 | 73 | |
AK131129.1 | - | 13 - 14 | 1775 | 73 | ||
AK143535.1 | - | 13 - 14 | 1815 | 73 | ||
AK149750.1 | - | 13 - 14 | 1816 | 73 | ||
AK166215.1 | - | 13 - 14 | 1815 | 73 | ||
AK169950.1 | - | 13 - 14 | 1811 | 73 | ||
AK171498.1 | - | 13 - 14 | 1858 | 73 | ||
BC059242.1 | - | 13 - 14 | 1813 | 73 | ||
BC060133.1 | - | 13 - 14 | 1813 | 73 | ||
BC079677.1 | - | 12 - 13 | 2481 | 73 | ||
BC141107.1 | - | 13 - 14 | 1781 | 9055719 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
