Assay Details
| Gene Symbol: Sox13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011439.2 | NP_035569.2 | 8 - 9 | 1178 | 79 | |
| GenBank mRNA | AB006329.1 | - | 7 - 8 | 834 | 79 | |
| AJ000740.1 | - | 7 - 8 | 785 | 79 | ||
| BC007130.1 | - | 8 - 9 | 1000 | 3493465 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sox13
Genomic Map