Assay Details
| Gene Symbol: Sprr2h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011474.3 | NP_035604.1 | 2 | 489 | 60 | |
| GenBank mRNA | AJ005566.1 | - | 1 | 461 | 60 | |
| AY158992.1 | - | 2 | 489 | 60 | ||
| BC099888.1 | - | 3 | 258 | 60 | ||
| BC099889.1 | - | 2 | 475 | 60 | ||
| BC099890.1 | - | 2 | 192 | 60 | ||
| BC127024.1 | - | 2 | 472 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P138462 | NM_011474.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P138462 | AJ005566.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P138462 | AY158992.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 386064 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P153992 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P134928 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P155841 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P138462 | NM_011474.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P138462 | AJ005566.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P138462 | AY158992.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22331176 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22331175 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22331185 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22331172 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22331188 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22331178 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22331180 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22331187 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22331142 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22331193 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22331177 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22331174 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22331182 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22331186 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P134928 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P153992 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P155841 | - | - |
| Whole Mouse Genome G4122A | A_51_P124665 | NM_011474.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P124665 | AJ005566.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P124665 | AY158992.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138462 | NM_011474.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138462 | AJ005566.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138462 | AY158992.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P155841 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P153992 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P134928 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02952870 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02952876 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02457653 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02457654 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02457649 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02952872 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02952877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331173 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331162 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331175 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331177 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331153 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331188 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02359482 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331180 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331142 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331183 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02801369 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331158 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03020859 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331155 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331169 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331204 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331200 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331147 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331173 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331193 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331178 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331185 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331195 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331167 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331192 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03045730 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331160 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331198 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02874773 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331150 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331202 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331186 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02457653 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331145 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02965003 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331164 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331170 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331182 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331176 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331163 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331187 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331166 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10493878 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493877 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422240_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10493876 | - | - |
| Mouse Genome 430 2.0 Array | 1422240_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1422240_PM_s_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20762_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20762_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300006409.mm.1 | NM_011474.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006409.mm.1 | NM_011474.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006409.mm.1 | TC0300000828.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300006405.mm.1 | NM_011474.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006405.mm.1 | NM_011474.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006405.mm.1 | TC0300000828.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300006408.mm.1 | NM_011474.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006408.mm.1 | NM_011474.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006408.mm.1 | TC0300000828.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300006410.mm.1 | NM_011474.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006410.mm.1 | NM_011474.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006410.mm.1 | TC0300000828.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300006411.mm.1 | NM_011474.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006411.mm.1 | NM_011474.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006411.mm.1 | TC0300000828.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300006407.mm.1 | NM_011474.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006407.mm.1 | NM_011474.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006407.mm.1 | TC0300000828.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1422240_s_at | - | - |
| Mouse Expression Set 430 | 1422240_PM_s_at | - | - |
| Murine Genome U74v2 Set | 94121_at | - | - |
| Murine Genome U74v2 Set | 101761_f_at | - | - |
| Murine Genome U74v2 Set | 101761_PM_f_at | - | - |
| Murine Genome U74v2 Set | 94121_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422240_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422240_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422240_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422240_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 597611 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2623615 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2623615 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2623615 | - | - |

Sprr2h
Genomic Map